{4-[5-(Trifluoromethyl)pyridin-2-yl]phenyl}methylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-[4-(Aminomethyl)phenyl]-5-(trifluoromethyl)pyridine, 4-[5-(Trifluoromethyl)pyridin-2-yl]benzylamine
PC8545-1G PC8545-250MG
APOSPC8545-1GEA 418 EUR
APOSPC8545-1G APOSPC8545-250MG
{4-[5-(Trifluoromethyl)pyridin-2-yl]phenyl}methylamine 97%
{4-[5-(Trifluoromethyl)pyridin-2-yl]phenyl}methylamine
Formula: C₁₃H₁₁F₃N₂
Melting Pt: 108.5…110 °C
Storage Temperature: Ambient
MDL Number: MFCD09817478
CAS Number: 906352-74-7
UN: 3259
ADR: 8,III