1-Bromo-1,1,2,2-tetrafluorobutane
Supplier: Apollo Scientific
Warning
PC8764-1G PC8764-5G
APOSPC8764-1GEA 58 EUR
APOSPC8764-1G APOSPC8764-5G
1-Bromo-1,1,2,2-tetrafluorobutane
1-Bromo-1,1,2,2-tetrafluorobutane
Formula: C₄H₅BrF₄
Boiling Pt: 143…144 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00077466
CAS Number: 127117-30-0