N-Methyl-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Supplier: Apollo Scientific
Synonyms: 2-[4-(N-Methylsulphamoyl)phenoxy]-3-(trifluoromethyl)pyridine
PC8877-1G PC8877-250MG
APOSPC8877-1GEA 270 EUR
APOSPC8877-1G APOSPC8877-250MG
N-Methyl-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N-Methyl-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formula: C₁₃H₁₁F₃N₂O₃S
MW: 332.298 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109527
CAS Number: 1227955-09-0