4-(2-Aminoethylthio)-8-fluoro-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
Warning
PC8886-1G PC8886-500MG
APOSPC8886-1GEA 248 EUR
APOSPC8886-1G APOSPC8886-500MG
4-(2-Aminoethylthio)-8-fluoro-2-(trifluoromethyl)quinoline 97%
4-(2-Aminoethylthio)-8-fluoro-2-(trifluoromethyl)quinoline
Formula: C₁₂H₁₀F₄N₂S
MW: 290.28 g/mol
Melting Pt: 102…103.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04038213
CAS Number: 541539-78-0