2-(2,4-Difluorophenyl)cyclopropane-1-carboxylic acid
Supplier: Apollo Scientific
Warning
PC8918-1G PC8918-250MG PC8918-500MG
APOSPC8918-1GEA 641 EUR
APOSPC8918-1G APOSPC8918-250MG APOSPC8918-500MG
2-(2,4-Difluorophenyl)cyclopropane-1-carboxylic acid
2-(2,4-Difluorophenyl)cyclopropane-1-carboxylic acid
Formula: C₁₀H₈F₂O₂
MW: 198.166 g/mol
Melting Pt: 307…310 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12068249
CAS Number: 1157642-22-2