3-(4-Fluorophenyl)propan-1-ol 96%
Supplier: Apollo Scientific
PC900029-1G PC900029-5G
APOSPC900029-1GEA 241 EUR
APOSPC900029-1G APOSPC900029-5G
3-(4-Fluorophenyl)propan-1-ol 96%
3-(4-Fluorophenyl)propan-1-ol
Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028733
CAS Number: 702-15-8