1-(Dimethoxymethyl)-2,4-difluorobenzene 98%
Supplier: Apollo Scientific
PC900128-10G PC900128-1G PC900128-5G
APOSPC900128-10GEA 502 EUR
APOSPC900128-10G APOSPC900128-1G APOSPC900128-5G
1-(Dimethoxymethyl)-2,4-difluorobenzene 98%
1-(Dimethoxymethyl)-2,4-difluorobenzene
Formula: C₉H₁₀F₂O₂
MW: 188.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22383753
CAS Number: 144062-35-1