4-Bromo-2-chloro-6-fluorobenzaldehyde 95%

Supplier: Apollo Scientific
Warning

PC900170-1G PC900170-250MG PC900170-5G
APOSPC900170-1GEA 316 EUR
APOSPC900170-1G APOSPC900170-250MG APOSPC900170-5G
4-Bromo-2-chloro-6-fluorobenzaldehyde 95%
4-Bromo-2-chloro-6-fluorobenzaldehyde
Formula: C₇H₃BrClFO
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD25460195
CAS Number: 929621-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR