5-Chloro-3-fluoro-2-methylpyridine 97%
Supplier: Apollo Scientific
PC900583-1G PC900583-250MG
APOSPC900583-1GEA 502 EUR
APOSPC900583-1G APOSPC900583-250MG
5-Chloro-3-fluoro-2-methylpyridine 97%
5-Chloro-3-fluoro-2-methylpyridine
Formula: C₆H₅ClFN
MW: 145.562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256290
CAS Number: 1210868-68-0