2-Chloro-5-(trifluoromethyl)pyridin-3-ol 98%
Supplier: Apollo Scientific
PC900598-1G PC900598-250MG
APOSPC900598-1GEA 299 EUR
APOSPC900598-1G APOSPC900598-250MG
2-Chloro-5-(trifluoromethyl)pyridin-3-ol 98%
2-Chloro-5-(trifluoromethyl)pyridin-3-ol
Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189916
CAS Number: 1196153-98-6