4-Fluoro-1-nitro-2-propoxybenzene 98%
Supplier: Apollo Scientific
PC901674-100G PC901674-25G PC901674-5G
APOSPC901674-100GEA 632 EUR
APOSPC901674-100G APOSPC901674-25G APOSPC901674-5G
4-Fluoro-1-nitro-2-propoxybenzene 98%
4-Fluoro-1-nitro-2-propoxybenzene
Formula: C₉H₁₀FNO₃
MW: 199.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19237155
CAS Number: 28987-45-3