1-(2,4-Difluorophenyl)piperidine 98%
Supplier: Apollo Scientific
PC902002-1G PC902002-5G
APOSPC902002-1GEA 145 EUR
APOSPC902002-1G APOSPC902002-5G
1-(2,4-Difluorophenyl)piperidine 98%
1-(2,4-Difluorophenyl)piperidine
Formula: C₁₁H₁₃F₂N
Storage Temperature: Ambient
MDL Number: MFCD20441851
CAS Number: 1345471-75-1