2-(3,4-Difluorophenyl)-2-methylpropanenitrile 98%
Supplier: Apollo Scientific
PC902018-1G PC902018-5G
APOSPC902018-1GEA 219 EUR
APOSPC902018-1G APOSPC902018-5G
2-(3,4-Difluorophenyl)-2-methylpropanenitrile 98%
2-(3,4-Difluorophenyl)-2-methylpropanenitrile
Formula: C₁₀H₉F₂N
Storage Temperature: Ambient
MDL Number: MFCD11036569
CAS Number: 1035262-16-8