1-(2-Fluorophenyl)-1H-pyrazole 98%
Supplier: Apollo Scientific
PC902185-1G PC902185-5G
APOSPC902185-1GEA 219 EUR
APOSPC902185-1G APOSPC902185-5G
1-(2-Fluorophenyl)-1H-pyrazole 98%
1-(2-Fluorophenyl)-1H-pyrazole
Formula: C₉H₇FN₂
MW: 162.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699187
CAS Number: 35715-66-3