1-Ethyl-6,7-difluoro-2-methyl-1,3-benzodiazole 98%
Supplier: Apollo Scientific
PC902288-1G PC902288-5G PC902288-10G
APOSPC902288-1GEA 168 EUR
APOSPC902288-1G APOSPC902288-5G APOSPC902288-10G
1-Ethyl-6,7-difluoro-2-methyl-1,3-benzodiazole 98%
1-Ethyl-6,7-difluoro-2-methyl-1,3-benzodiazole
Formula: C₁₀H₁₀F₂N₂
MW: 196.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22375041
CAS Number: 1381944-71-3