(S)-1-(4-Bromo-2-fluorophenyl)ethanamine hydrochloride 95%
Supplier: Apollo Scientific
PC902865-1G PC902865-250MG
APOSPC902865-1GEA 328 EUR
APOSPC902865-1G APOSPC902865-250MG
(S)-1-(4-Bromo-2-fluorophenyl)ethanamine hydrochloride 95%
(S)-1-(4-Bromo-2-fluorophenyl)ethanamine hydrochloride
Formula: C₈H₉BrFN·ClH
MW: 254.527 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19382355
CAS Number: 1311254-85-9