5-Hydroxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
PC904015-1G PC904015-5G
APOSPC904015-1GEA 102 EUR
APOSPC904015-1G APOSPC904015-5G
5-Hydroxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 95%
5-Hydroxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formula: C₆H₅F₃N₂O₂
MW: 194.111 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12827694
CAS Number: 179732-64-0