2-Fluoro-6-(methylsulphonyl)toluene 95%
Supplier: Apollo Scientific
PC904515-1G PC904515-250MG
APOSPC904515-1GEA 734 EUR
APOSPC904515-1G APOSPC904515-250MG
2-Fluoro-6-(methylsulphonyl)toluene 95%
2-Fluoro-6-(methylsulphonyl)toluene
Formula: C₈H₉FO₂S
MW: 188.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527256
CAS Number: 828270-59-3