(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride 97%
Supplier: Apollo Scientific
Warning
PC904802-1G PC904802-250MG
APOSPC904802-1GEA 31.9 EUR
APOSPC904802-1G APOSPC904802-250MG
(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride 97%
(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride
Formula: C₈H₁₀FNO·ClH
MW: 191.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546099
CAS Number: 1214366-31-0