6-Fluoro-[1,2,4]triazolo[1,5-a]pyridin-2-amine 95%
Supplier: Apollo Scientific
PC904990-1G PC904990-250MG
APOSPC904990-1GEA 806 EUR
APOSPC904990-1G APOSPC904990-250MG
6-Fluoro-[1,2,4]triazolo[1,5-a]pyridin-2-amine 95%
6-Fluoro-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula: C₆H₅FN₄
MW: 152.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072710
CAS Number: 1245644-40-9