6-Fluoro-2-methoxynicotinonitrile 95%

Supplier: Apollo Scientific
Warning

PC905158-1G PC905158-250MG
APOSPC905158-1GEA 394 EUR
APOSPC905158-1G APOSPC905158-250MG
6-Fluoro-2-methoxynicotinonitrile 95%
6-Fluoro-2-methoxynicotinonitrile
Formula: C₇H₅FN₂O
MW: 152.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26743428
CAS Number: 1339175-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR