4-Aminomethyl-2,2-difluoro-1,3-benzodioxole 96%
Supplier: Apollo Scientific
PC905533-1G PC905533-250MG
APOSPC905533-1GEA 125 EUR
APOSPC905533-1G APOSPC905533-250MG
4-Aminomethyl-2,2-difluoro-1,3-benzodioxole 96%
4-Aminomethyl-2,2-difluoro-1,3-benzodioxole
Formula: C₈H₇F₂NO₂
MW: 187.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16617616
CAS Number: 531508-46-0