2,2-Difluoropropylamine hydrochloride 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Amino-2,2-difluoropropane hydrochloride, 2,2-Difluoropropan-1-amine hydrochloride
PC9057-1G PC9057-250MG PC9057-5G
APOSPC9057-1GEA 27.7 EUR
APOSPC9057-1G APOSPC9057-250MG APOSPC9057-5G
2,2-Difluoropropylamine hydrochloride 95+%
2,2-Difluoropropylamine hydrochloride
Formula: C₃H₇F₂N·ClH
MW: 131.552 g/mol
Melting Pt: 235…239 °C
Density: 0.936 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06659051
CAS Number: 868241-48-9