3-(Trifluoromethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride ≥98%
Supplier: Apollo Scientific
PC907901-1G PC907901-250MG
APOSPC907901-1GEA 1540 EUR
APOSPC907901-1G APOSPC907901-250MG
3-(Trifluoromethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride ≥98%
3-(Trifluoromethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride
Formula: C₇H₈F₃N₃·ClH
MW: 227.615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443824
CAS Number: 733757-88-5