1-(3,4-Difluorophenyl)ethanol 98%
Supplier: Apollo Scientific
Warning
PC908932-1G PC908932-5G
APOSPC908932-1GEA 21.9 EUR
APOSPC908932-1G APOSPC908932-5G
1-(3,4-Difluorophenyl)ethanol 98%
1-(3,4-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08442288
CAS Number: 321318-21-2