2-Methyl-4-(trifluoromethyl)benzaldehyde 95%
Supplier: Apollo Scientific
PC909109-1G PC909109-5G
APOSPC909109-1GEA 119 EUR
APOSPC909109-1G APOSPC909109-5G
2-Methyl-4-(trifluoromethyl)benzaldehyde 95%
2-Methyl-4-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
MW: 188.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399853
CAS Number: 888739-67-1