4-(3-Fluorophenyl)phenol 97%

Supplier: Apollo Scientific

PC909235-1G PC909235-5G
APOSPC909235-1GEA 219 EUR
APOSPC909235-1G APOSPC909235-5G
4-(3-Fluorophenyl)phenol 97%
4-(3-Fluorophenyl)phenol
Formula: C₁₂H₉FO
MW: 188.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802291
CAS Number: 64465-61-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR