1,2-Difluoro-4-methoxy-5-nitrobenzene 95%
Supplier: Apollo Scientific
Warning
PC909902-1G PC909902-250MG
APOSPC909902-1GEA 140 EUR
APOSPC909902-1G APOSPC909902-250MG
1,2-Difluoro-4-methoxy-5-nitrobenzene 95%
1,2-Difluoro-4-methoxy-5-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040445
CAS Number: 66684-64-8