2,2-Difluoro-1,3-benzodioxol-4-ol 95%
Supplier: Apollo Scientific
PC910274-1G PC910274-250MG
APOSPC910274-1GEA 364 EUR
APOSPC910274-1G APOSPC910274-250MG
2,2-Difluoro-1,3-benzodioxol-4-ol 95%
2,2-Difluoro-1,3-benzodioxol-4-ol
Formula: C₇H₄F₂O₃
MW: 174.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18451422
CAS Number: 126120-86-3