(R)-5-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 97%
Supplier: Apollo Scientific
Warning
PC910470-100MG PC910470-250MG
APOSPC910470-100MGEA 82.8 EUR
APOSPC910470-100MG APOSPC910470-250MG
(R)-5-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 97%
(R)-5-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride
Formula: C₉H₁₀FN·ClH
MW: 187.642 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295031
CAS Number: 1381928-19-3