5-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid 95%
Supplier: Apollo Scientific
PC910578-1G PC910578-250MG
APOSPC910578-1GEA 492 EUR
APOSPC910578-1G APOSPC910578-250MG
5-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid 95%
5-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid
Formula: C₆H₆F₂N₂O₂
MW: 176.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827721
CAS Number: 1204298-65-6