7-Fluoro-6-methoxy-3H-quinazolin-4-one 96%
Supplier: Apollo Scientific
PC910741-1G PC910741-250MG
APOSPC910741-1GEA 940 EUR
APOSPC910741-1G APOSPC910741-250MG
7-Fluoro-6-methoxy-3H-quinazolin-4-one 96%
7-Fluoro-6-methoxy-3H-quinazolin-4-one
Formula: C₉H₇FN₂O₂
MW: 194.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23105805
CAS Number: 869475-52-5