4-(4-Fluorophenyl)-2-methylthiazole 98%
Supplier: Apollo Scientific
PC910856-1G
PC910856-5G
APOSPC910856-1GEA
168
EUR
APOSPC910856-1G
APOSPC910856-5G
4-(4-Fluorophenyl)-2-methylthiazole 98%
4-(4-Fluorophenyl)-2-methylthiazole
Formula:
C₁₀H₈FNS MW: 193.241 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00518914 CAS Number: 450-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...