6-Fluoro-4-methyl-2H-1,4-benzoxazin-3-one 98%
Supplier: Apollo Scientific
PC910923-1G PC910923-250MG
APOSPC910923-1GEA 676 EUR
APOSPC910923-1G APOSPC910923-250MG
6-Fluoro-4-methyl-2H-1,4-benzoxazin-3-one 98%
6-Fluoro-4-methyl-2H-1,4-benzoxazin-3-one
Formula: C₉H₈FNO₂
MW: 181.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405626
CAS Number: 1260796-58-4