2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 97%
Supplier: Apollo Scientific
PC910944-1G PC910944-250MG
APOSPC910944-1GEA 377 EUR
APOSPC910944-1G APOSPC910944-250MG
2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 97%
2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine
Formula: C₇H₄F₃N₃
MW: 187.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19705789
CAS Number: 73221-25-7