2-(2-Fluorophenyl)propan-2-amine hydrochloride 98%
Supplier: Apollo Scientific
Warning
PC911159-1G PC911159-5G
APOSPC911159-1GEA 208 EUR
APOSPC911159-1G APOSPC911159-5G
2-(2-Fluorophenyl)propan-2-amine hydrochloride 98%
2-(2-Fluorophenyl)propan-2-amine hydrochloride
Formula: C₉H₁₂FN·ClH
MW: 189.658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870090
CAS Number: 1202751-82-3