1-(2,3-Difluorophenyl)ethanol 95%
Supplier: Apollo Scientific
PC911367-1G PC911367-5G
APOSPC911367-1GEA 190 EUR
APOSPC911367-1G APOSPC911367-5G
1-(2,3-Difluorophenyl)ethanol 95%
1-(2,3-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18911748
CAS Number: 1228690-56-9