4-Amino-2,3-difluorobenzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 4-Cyano-2,3-difluoroaniline
PC9170-1G PC9170-5G
APOSPC9170-1GEA 40.6 EUR
APOSPC9170-1G APOSPC9170-5G
4-Amino-2,3-difluorobenzonitrile
4-Amino-2,3-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154.117 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09030642
CAS Number: 112279-71-7
UN: 3439
ADR: 6.1 III