4-Bromo-2,3-difluorobenzonitrile 95%
Supplier: Apollo Scientific
Warning
PC9171-1G PC9171-5G PC9171-250MG
APOSPC9171-1GEA 45.1 EUR
APOSPC9171-1G APOSPC9171-5G APOSPC9171-250MG
4-Bromo-2,3-difluorobenzonitrile 95%
4-Bromo-2,3-difluorobenzonitrile
Formula: C₇H₂BrF₂N
MW: 217.998 g/mol
Melting Pt: 51…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09030640
CAS Number: 126163-58-4