2-(1,1,2,3,3,3-Hexafluoropropyl)tetrahydrofuran 95%
Supplier: Apollo Scientific
Warning
PC9224-25G PC9224-5G
APOSPC9224-25GEA 607 EUR
APOSPC9224-25G APOSPC9224-5G
2-(1,1,2,3,3,3-Hexafluoropropyl)tetrahydrofuran 95%
2-(1,1,2,3,3,3-Hexafluoropropyl)tetrahydrofuran
Formula: C₇H₈F₆O
MW: 222.128 g/mol
Boiling Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00155855
CAS Number: 53005-42-8
UN: 1993
ADR: 3,III