2-Fluoro-3-methoxybenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Chlorocarbonyl)-2-fluoroanisole
PC9296-1G PC9296-25G PC9296-5G
APOSPC9296-1GEA 65.4 EUR
APOSPC9296-1G APOSPC9296-25G APOSPC9296-5G
2-Fluoro-3-methoxybenzoyl chloride 97%
2-Fluoro-3-methoxybenzoyl chloride
Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Boiling Pt: 133 °C (12 mm Hg)
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD08277634
CAS Number: 850563-45-0
UN: 3261
ADR: 8 III