2-Chloro-3-(trifluoromethyl)benzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-3-(chlorocarbonyl)benzotrifluoride
PC9297-1G PC9297-25G
APOSPC9297-1GEA 29 EUR
APOSPC9297-1G APOSPC9297-25G
2-Chloro-3-(trifluoromethyl)benzoyl chloride 98%
2-Chloro-3-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Boiling Pt: 224 °C
Melting Pt: 33…35 °C
Storage Temperature: Ambient
MDL Number: MFCD04115830
CAS Number: 850156-39-7
UN: 3261
ADR: 8,III