2,4-Difluorophenylacetyl chloride 99%
Supplier: Apollo Scientific
Danger
PC9369-1G PC9369-5G
APOSPC9369-1GEA 49.3 EUR
APOSPC9369-1G APOSPC9369-5G
2,4-Difluorophenylacetyl chloride 99%
2,4-Difluorophenylacetyl chloride
Formula: C₈H₅ClF₂O
MW: 190.575 g/mol
Boiling Pt: 85…87 °C (11 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08458139
CAS Number: 141060-00-6
UN: 1760
ADR: 8,III