2-(2,5-Difluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
Supplier: Apollo Scientific
|
Warning
|
PC9557-1G
PC9557-5G
APOSPC9557-1GEA
256
EUR
APOSPC9557-1G
APOSPC9557-5G
2-(2,5-Difluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
2-(2,5-Difluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide
Formula:
C₁₅H₁₀F₂N₂OS·BrH Melting Pt: >230 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458147 CAS Number: 1210224-35-3 |