1-Bromo-1-fluoroethene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Bromo-1-fluoroethylene
PC9606-25G PC9606-5G
APOSPC9606-25GEA 720 EUR
APOSPC9606-25G APOSPC9606-5G
1-Bromo-1-fluoroethene 97%
1-Bromo-1-fluoroethene
Formula: C₂H₂BrF
MW: 124.94 g/mol
Boiling Pt: 6.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04038866
CAS Number: 420-25-7
UN: 3161
ADR: 2.1,