3,3,3-Trifluoro-2-(trifluoromethyl)-1,2-propenoxide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Epoxy-1,1,1-trifluoro-2-(trifluoromethyl)propane, 2,2-Bis(trifluoromethyl)oxirane
PC9626-25G PC9626-1G PC9626-5G
APOSPC9626-25GEA 906 EUR
APOSPC9626-25G APOSPC9626-1G APOSPC9626-5G
3,3,3-Trifluoro-2-(trifluoromethyl)-1,2-propenoxide 97%
3,3,3-Trifluoro-2-(trifluoromethyl)-1,2-propenoxide
Formula: C₄H₂F₆O
Boiling Pt: 41…42 °C
Melting Pt: –112 °C
Density: 1.493 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD07784232
CAS Number: 31898-68-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM