3,5-Bis(trifluoromethyl)-α-methylbenzylamine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine
PC9791-1G PC9791-5G
APOSPC9791-1GEA 364 EUR
APOSPC9791-1G APOSPC9791-5G
3,5-Bis(trifluoromethyl)-α-methylbenzylamine 95%
3,5-Bis(trifluoromethyl)-α-methylbenzylamine
Formula: C₁₀H₉F₆N
MW: 257.176 g/mol
Boiling Pt: 6.5…64.5 °C (3.2 mm Hg)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD04038875
CAS Number: 187085-97-8