2-(4-Bromo-2,6-difluorophenyl)-2-methyl-1,3-dioxolane 97%
Supplier: Apollo Scientific
PC99028-1G PC99028-500MG PC99028-5G
APOSPC99028-1GEA 188 EUR
APOSPC99028-1G APOSPC99028-500MG APOSPC99028-5G
2-(4-Bromo-2,6-difluorophenyl)-2-methyl-1,3-dioxolane 97%
2-(4-Bromo-2,6-difluorophenyl)-2-methyl-1,3-dioxolane
Formula: C₁₀H₉BrF₂O₂
MW: 279.078 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707280
CAS Number: 2484889-09-8