2-Fluoro-6-iodonitrobenzene 96%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-3-iodo-2-nitrobenzene
PC9920-1G PC9920-5G PC9920-10G
APOSPC9920-1GEA 31.9 EUR
APOSPC9920-1G APOSPC9920-5G APOSPC9920-10G
2-Fluoro-6-iodonitrobenzene 96%
2-Fluoro-6-iodonitrobenzene
Formula: C₆H₃FINO₂
MW: 266.996 g/mol
Melting Pt: 36…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368790
CAS Number: 886762-71-6