4-(4-Fluorophenoxy)benzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
PC9934-1G PC9934-250MG
APOSPC9934-1GEA 400 EUR
APOSPC9934-1G APOSPC9934-250MG
4-(4-Fluorophenoxy)benzenesulphonyl chloride 95%
4-(4-Fluorophenoxy)benzenesulphonyl chloride
Formula: C₁₂H₈ClFO₃S
MW: 286.707 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631868
CAS Number: 192329-91-2
UN: 3261
ADR: 8,III